Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hnrnpl
Entrez Gene ID
394074
Gene Name
heterogeneous nuclear ribonucleoprotein L
Gene Aliases
hnrpl, zgc:55429
Chromosome Location
Chr.5: 141172 - 156962 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201099.1 NP_957393.1 10 - 11 1219 96
XM_021476043.1 XP_021331718.1 9 - 10 1205 96
GenBank mRNA BC045336.1 - 10 - 11 1219 96
BC164693.1 - 10 - 11 1231 96

View Full Product Details