Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lyrm1
Entrez Gene ID
436779
Gene Name
LYR motif containing 1
Gene Aliases
wu:fj36g10, zgc:92830
Chromosome Location
Chr.24: 39510735 - 39518618 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002506.1 NP_001002506.1 3 - 4 294 103
GenBank mRNA BC076293.1 - 3 - 4 294 103

View Full Product Details