Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcdhb
Entrez Gene ID
450003
Gene Name
glutaryl-CoA dehydrogenase b
Gene Aliases
gcdhl, zgc:103477
Chromosome Location
Chr.3: 13941010 - 13955886 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006024.1 NP_001006024.1 1 - 2 193 106
GenBank mRNA BC083397.1 - 1 - 2 193 106
BC164011.1 - 1 - 2 167 106

View Full Product Details