Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smyd2a
Entrez Gene ID
541423
Gene Name
SET and MYND domain containing 2a
Gene Aliases
im:7146196, smyd2, wu:fc02f02, zgc:110553
Chromosome Location
Chr.17: 32745822 - 32779439 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013550.1 NP_001013568.1 4 - 5 546 86
GenBank mRNA BC091465.1 - 4 - 5 546 86
DQ851823.1 - 4 - 5 429 86

View Full Product Details