Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smyd2a
Entrez Gene ID
541423
Gene Name
SET and MYND domain containing 2a
Gene Aliases
im:7146196, smyd2, wu:fc02f02, zgc:110553
Chromosome Location
Chr.17: 32745822 - 32779439 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013550.1 NP_001013568.1 6 - 7 745 67
GenBank mRNA BC091465.1 - 6 - 7 745 67

View Full Product Details