Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gnsa
Entrez Gene ID
566506
Gene Name
glucosamine (N-acetyl)-6-sulfatase a
Gene Aliases
zgc:114066
Chromosome Location
Chr.4: 4510057 - 4524280 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030208.1 NP_001025379.1 12 - 13 1517 100
GenBank mRNA BC097128.1 - 12 - 13 1517 100

View Full Product Details