The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077334.2 | NP_001070802.1 | 3 - 4 | 847 | 77 |
GenBank mRNA | BC124624.1 | - | 3 - 4 | 848 | 77 |
BC165277.1 | - | 3 - 4 | 705 | 77 | |
GU434294.1 | - | 3 - 4 | 671 | 77 |