Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ipmkb
Entrez Gene ID
790916
Gene Name
inositol polyphosphate multikinase b
Gene Aliases
zgc:158331
Chromosome Location
Chr.12: 7198761 - 7234915 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080064.1 NP_001073533.1 1 - 2 330 68
GenBank mRNA BC128841.1 - 1 - 2 330 68

View Full Product Details