Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prkra
Entrez Gene ID
557370
Gene Name
protein kinase, interferon-inducible double stranded RNA dependent activator
Gene Aliases
im:7151758, zgc:162619
Chromosome Location
Chr.9: 1365728 - 1371542 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089361.1 NP_001082830.1 2 - 3 252 71
XM_005172508.4 XP_005172565.1 2 - 3 236 71
GenBank mRNA BC135034.1 - 2 - 3 252 71

View Full Product Details