Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stxbp1b
Entrez Gene ID
557717
Gene Name
syntaxin binding protein 1b
Gene Aliases
si:rp71-10d23.3
Chromosome Location
Chr.5: 60754 - 87599 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089376.1 NP_001082845.1 10 - 11 936 81
GenBank mRNA BC134211.1 - 10 - 11 929 81
BC171552.1 - 10 - 11 918 81

View Full Product Details