Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stxbp1b
Entrez Gene ID
557717
Gene Name
syntaxin binding protein 1b
Gene Aliases
si:rp71-10d23.3
Chromosome Location
Chr.5: 60754 - 87599 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089376.1 NP_001082845.1 5 - 6 362 117
GenBank mRNA BC134211.1 - 5 - 6 355 117
BC171526.1 - 5 - 6 344 117
BC171552.1 - 5 - 6 344 117

View Full Product Details