Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thbs4b
Entrez Gene ID
252850
Gene Name
thrombospondin 4b
Gene Aliases
thbs4, tsp4, zgc:111910
Chromosome Location
Chr.21: 5129523 - 5162687 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173226.1 NP_775333.1 21 - 22 2927 86
GenBank mRNA AY081009.1 - 21 - 22 2927 86
BC095593.1 - 21 - 22 2937 86

View Full Product Details