Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zc4h2
Entrez Gene ID
323487
Gene Name
zinc finger, C4H2 domain containing
Gene Aliases
wu:fb99f08, zgc:63849
Chromosome Location
Chr.5: 65992 - 75111 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199642.1 NP_955936.1 4 - 5 669 81
GenBank mRNA AY648796.1 - 4 - 5 696 81
BC053110.1 - 4 - 5 669 81

View Full Product Details