Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndrg3b
Entrez Gene ID
327395
Gene Name
ndrg family member 3b
Gene Aliases
NDRG3, fi05h10, wu:fi05h10, zgc:55301
Chromosome Location
Chr.23: 14832540 - 14990916 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199797.1 NP_956091.1 14 - 15 1007 68
XM_005161986.2 XP_005162043.1 14 - 15 1101 68
XM_009296689.1 XP_009294964.1 14 - 15 1101 68
XM_009296690.2 XP_009294965.1 12 - 13 909 68
XM_009296691.2 XP_009294966.1 14 - 15 1101 68
XM_017353522.1 XP_017209011.1 14 - 15 1101 68
GenBank mRNA BC044139.1 - 14 - 15 1007 68

View Full Product Details