Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
foxk1
Entrez Gene ID
334470
Gene Name
forkhead box K1
Gene Aliases
wu:fi75f05, zgc:66237
Chromosome Location
Chr.3: 40902060 - 40932962 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199902.1 NP_956196.1 7 - 8 1576 65
GenBank mRNA BC054664.1 - 7 - 8 1576 65

View Full Product Details