Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bcap31
Entrez Gene ID
368488
Gene Name
B cell receptor associated protein 31
Gene Aliases
si:bz30i22.4, si:rp71-30i22.4, zgc:56389
Chromosome Location
Chr.8: 8973425 - 9029176 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200092.1 NP_956386.1 5 - 6 580 89
GenBank mRNA BC049014.1 - 5 - 6 580 89

View Full Product Details