Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
waslb
Entrez Gene ID
394034
Gene Name
WASP like actin nucleation promoting factor b
Gene Aliases
wasl, zgc:55478, zgc:77237
Chromosome Location
Chr.4: 4071138 - 4111771 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201059.2 NP_957353.2 3 - 4 532 126
XM_005164482.4 XP_005164539.1 3 - 4 613 126
XM_021474971.1 XP_021330646.1 3 - 4 308 126
GenBank mRNA BC044180.1 - 3 - 4 582 126
BC066424.1 - 3 - 4 532 126

View Full Product Details