Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grhprb
Entrez Gene ID
402985
Gene Name
glyoxylate reductase/hydroxypyruvate reductase b
Gene Aliases
fb80f04, wu:fb80f04, zgc:77636
Chromosome Location
Chr.1: 17050 - 26780 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205685.2 NP_991248.2 2 - 3 494 93
XM_005168340.4 XP_005168397.1 2 - 3 489 93
GenBank mRNA BC052108.1 - 2 - 3 333 93
BC065431.1 - 2 - 3 354 93

View Full Product Details