Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccni
Entrez Gene ID
406239
Gene Name
cyclin I
Gene Aliases
im:4967226, wu:fb18c12, wu:fk71g05, zgc:158424, zgc:55449, zgc:85721
Chromosome Location
Chr.21: 5780886 - 5799122 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213221.1 NP_998386.1 4 - 5 560 70
GenBank mRNA BC044400.1 - 4 - 5 548 70
BC068369.1 - 4 - 5 560 70

View Full Product Details