The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214722.1 | NP_999887.1 | 2 - 3 | 396 | 94 |
GenBank mRNA | BC064286.1 | - | 2 - 3 | 396 | 94 |