Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thrap3b
Entrez Gene ID
407984
Gene Name
thyroid hormone receptor associated protein 3b
Gene Aliases
zgc:85716
Chromosome Location
Chr.19: 3865182 - 3887183 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214815.1 NP_999980.1 10 - 11 2630 105
XM_005173742.4 XP_005173799.1 11 - 12 2609 105
XM_021468043.1 XP_021323718.1 10 - 11 2742 105
XM_021468044.1 XP_021323719.1 8 - 9 1670 105
XM_021468045.1 XP_021323720.1 10 - 11 1918 105
GenBank mRNA BC070004.1 - 10 - 11 2630 105

View Full Product Details