Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hvcn1
Entrez Gene ID
436618
Gene Name
hydrogen voltage-gated channel 1
Gene Aliases
zgc:92181
Chromosome Location
Chr.10: 8402070 - 8412964 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002346.1 NP_001002346.1 5 - 6 724 92
GenBank mRNA BC075916.1 - 5 - 6 724 92

View Full Product Details