Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsdl1
Entrez Gene ID
494064
Gene Name
hydroxysteroid dehydrogenase like 1
Gene Aliases
si:ch211-172b19.1, zgc:103498
Chromosome Location
Chr.20: 40761726 - 40766387 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008607.1 NP_001008607.1 3 - 4 1077 70
GenBank mRNA BC086821.1 - 3 - 4 1077 70

View Full Product Details