Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fgf19
Entrez Gene ID
368245
Gene Name
fibroblast growth factor 19
Gene Aliases
-
Chromosome Location
Chr.7: 54641996 - 54645443 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012246.2 NP_001012246.2 2 - 3 403 92
NM_001309359.1 NP_001296288.1 2 - 3 399 92
GenBank mRNA AB183515.1 - 2 - 3 307 92
BC162870.1 - 2 - 3 405 92
EH498287.1 - 2 - 3 420 92

View Full Product Details