Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nfyba
Entrez Gene ID
550227
Gene Name
nuclear transcription factor Y, beta a
Gene Aliases
zgc:110552
Chromosome Location
Chr.4: 9178007 - 9183605 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017565.1 NP_001017565.1 6 - 7 609 85
XM_005168523.4 XP_005168580.1 6 - 7 605 85
GenBank mRNA BC092926.1 - 6 - 7 609 85

View Full Product Details