Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
capslb
Entrez Gene ID
550473
Gene Name
calcyphosine-like b
Gene Aliases
zgc:110369
Chromosome Location
Chr.21: 22423330 - 22428328 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017776.2 NP_001017776.2 3 - 4 392 75
GenBank mRNA BC092903.1 - 3 - 4 393 75

View Full Product Details