Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnnt2d
Entrez Gene ID
574000
Gene Name
troponin T2d, cardiac
Gene Aliases
zgc:114184
Chromosome Location
Chr.25: 20215983 - 20226584 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025179.1 NP_001020350.1 6 - 7 293 60
GenBank mRNA BC097233.1 - 6 - 7 293 60

View Full Product Details