Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnmt3bb.1
Entrez Gene ID
317744
Gene Name
DNA (cytosine-5-)-methyltransferase 3 beta, duplicate b.1
Gene Aliases
cb633, dnmt3b, dnmt4
Chromosome Location
Chr.23: 17010893 - 17101407 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025450.1 NP_001020621.1 15 - 16 1718 75
XM_005162028.4 XP_005162085.1 15 - 16 1922 75
XM_009296716.3 XP_009294991.1 14 - 15 1776 75
XM_009296717.3 XP_009294992.1 15 - 16 1926 75
GenBank mRNA AB196915.1 - 15 - 16 1718 75
BC124098.1 - 15 - 16 1802 75

View Full Product Details