Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc17a8
Entrez Gene ID
563467
Gene Name
solute carrier family 17 member 8
Gene Aliases
si:ch211-264e16.4, vglut3
Chromosome Location
Chr.18: 158044 - 176551 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082835.1 NP_001076304.1 6 - 7 766 80

View Full Product Details