Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc22a7a
Entrez Gene ID
794188
Gene Name
solute carrier family 22 member 7a
Gene Aliases
im:6901703, im:7155670, oat2a, zgc:162334
Chromosome Location
Chr.11: 30729745 - 30742966 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083861.1 NP_001077330.1 7 - 8 1391 89
GenBank mRNA BC134964.1 - 7 - 8 1391 89

View Full Product Details