Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hadhab
Entrez Gene ID
793834
Gene Name
hydroxyacyl-CoA dehydrogenase trifunctional multienzyme complex subunit alpha b
Gene Aliases
im:7144234, zgc:158138
Chromosome Location
Chr.17: 37227961 - 37252810 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089437.1 NP_001082906.1 16 - 17 1699 86
GenBank mRNA BC096816.1 - 16 - 17 1724 86
BC131856.1 - 16 - 17 1699 86

View Full Product Details