Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hirip3
Entrez Gene ID
100000570
Gene Name
HIRA interacting protein 3
Gene Aliases
fi16e03, wu:fi16e03, zgc:66480
Chromosome Location
Chr.3: 119220 - 136200 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199823.1 NP_956117.1 1 - 2 107 63
GenBank mRNA BC055674.1 - 1 - 2 107 63

View Full Product Details