Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pes
Entrez Gene ID
30228
Gene Name
pescadillo
Gene Aliases
-
Chromosome Location
Chr.5: 3604 - 23843 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131030.3 NP_571105.3 4 - 5 501 66
GenBank mRNA BC049339.1 - 4 - 5 66
BC160624.1 - 4 - 5 407 66
U77627.1 - 4 - 5 467 66

View Full Product Details