Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acat2
Entrez Gene ID
30643
Gene Name
acetyl-CoA acetyltransferase 2
Gene Aliases
fb10a06, fb53f08, wu:fb10a06, wu:fb53f08
Chromosome Location
Chr.20: 30922595 - 30931182 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131370.2 NP_571445.2 2 - 3 262 120
XM_021468355.1 XP_021324030.1 2 - 3 305 120
GenBank mRNA AF143488.1 - 2 - 3 207 120
BC045949.1 - 2 - 3 262 120
BC165310.1 - 2 - 3 219 120

View Full Product Details