Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- trpc4apa
- Entrez Gene ID
-
58090
- Gene Name
- transient receptor potential cation channel, subfamily C, member 4 associated protein a
- Gene Aliases
-
tap1a, wu:fb15d09, zgc:109818
- Chromosome Location
-
Chr.11: 25157375 - 25180330 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_131569.2 |
NP_571644.1 |
6 - 7 |
616 |
137 |
GenBank mRNA |
AF130459.1 |
- |
6 - 7 |
617 |
137 |
View Full Product Details