Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pbx2
Entrez Gene ID
58136
Gene Name
pre-B-cell leukemia homeobox 2
Gene Aliases
pbxy, wu:fb73g03
Chromosome Location
Chr.16: 48616220 - 48643191 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131612.2 NP_571687.1 2 - 3 410 71
XM_017351360.2 XP_017206849.1 2 - 3 401 71
GenBank mRNA AF210642.1 - 1 - 2 71
AJ245965.1 - - 71
BC162623.1 - 2 - 3 410 71

View Full Product Details