Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scn8aa
Entrez Gene ID
58152
Gene Name
sodium channel, voltage gated, type VIII, alpha subunit a
Gene Aliases
Nav1.6, scn8a
Chromosome Location
Chr.23: 27248689 - 27345450 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131628.1 NP_571703.1 9 - 10 1422 97
XM_005162268.4 XP_005162325.1 9 - 10 1433 97
XM_009296871.3 XP_009295146.1 9 - 10 1433 97
XM_009296874.3 XP_009295149.1 9 - 10 1432 97
XM_021469841.1 XP_021325516.1 9 - 10 1578 97
XM_021469842.1 XP_021325517.1 9 - 10 1447 97
XM_021469843.1 XP_021325518.1 9 - 10 1446 97
GenBank mRNA AF297658.1 - 9 - 10 1422 97
BC163773.1 - 9 - 10 1243 97

View Full Product Details