Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snapc1a
Entrez Gene ID
393233
Gene Name
small nuclear RNA activating complex, polypeptide 1a
Gene Aliases
snapc1, zgc:56450
Chromosome Location
Chr.20: 20314579 - 20325492 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200263.1 NP_956557.1 3 - 4 369 66
XM_021468375.1 XP_021324050.1 2 - 3 1169 66
XM_021468376.1 XP_021324051.1 3 - 4 1094 66
GenBank mRNA BC049022.1 - 3 - 4 369 66

View Full Product Details