Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- atp6v0cb
- Entrez Gene ID
-
325402
- Gene Name
- ATPase H+ transporting V0 subunit cb
- Gene Aliases
-
wu:fc74d11, zgc:111904, zgc:77708
- Chromosome Location
-
Chr.24: 39158283 - 39175750 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205554.1 |
NP_991117.1 |
1 - 2 |
248 |
103 |
GenBank mRNA |
BC065849.1 |
- |
1 - 2 |
248 |
103 |
- Gene Symbol
- camta2
- Entrez Gene ID
-
101886599
- Gene Name
- calmodulin binding transcription activator 2
- Gene Aliases
-
si:ch1073-98b4.2
- Chromosome Location
-
Chr.23: 44655867 - 44706454 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BM157242.1 |
- |
- |
267 |
103 |
View Full Product Details