Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eif4h
Entrez Gene ID
402996
Gene Name
eukaryotic translation initiation factor 4h
Gene Aliases
wbscr1, zgc:77282
Chromosome Location
Chr.15: 4585401 - 4616819 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197328.1 NP_001184257.1 1 - 2 140 91
NM_001197329.1 NP_001184258.1 1 - 2 140 91
NM_205695.2 NP_991258.2 1 - 2 140 91
XM_009291449.3 XP_009289724.1 1 - 2 143 91
GenBank mRNA BC056285.1 - 1 - 2 82 91
BC065640.1 - 1 - 2 85 91
CT633866.2 - 1 - 2 66 91
EH552521.1 - 1 - 2 133 91
FP141411.1 - 1 - 2 117 91

View Full Product Details