Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdk7
Entrez Gene ID
405897
Gene Name
cyclin-dependent kinase 7
Gene Aliases
zgc:85821
Chromosome Location
Chr.5: 52000159 - 52010153 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212961.2 NP_998126.2 2 - 3 190 68
GenBank mRNA BC097024.1 - 2 - 3 172 68
DQ294347.1 - 2 - 3 190 68

View Full Product Details