Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eif3i
Entrez Gene ID
406263
Gene Name
eukaryotic translation initiation factor 3, subunit I
Gene Aliases
eIF3 p36, eif3s2, hm:zehn0464, wu:fb62b08, zgc:56211, zgc:77823
Chromosome Location
Chr.19: 29822500 - 29832876 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212990.1 NP_998155.1 8 - 9 883 103
GenBank mRNA BC045995.1 - 8 - 9 883 103
BC065874.1 - 8 - 9 825 103

View Full Product Details