Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgrrf1
Entrez Gene ID
692284
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
si:dkey-29m11.5, si:dkey-63j12.2, zgc:136229
Chromosome Location
Chr.20: 148602 - 152500 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045257.1 NP_001038722.1 1 - 2 206 130
GenBank mRNA BC116465.1 - 1 - 2 206 130

View Full Product Details