Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
agpat9l
Entrez Gene ID
567414
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 9, like
Gene Aliases
si:ch211-155m12.4
Chromosome Location
Chr.5: 10420 - 47370 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099450.1 NP_001092920.1 8 - 9 992 81

View Full Product Details