Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prdm8b
Entrez Gene ID
557162
Gene Name
PR domain containing 8b
Gene Aliases
si:ch211-57h10.4
Chromosome Location
Chr.5: 133919 - 138735 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105105.1 NP_001098575.1 3 - 4 647 75

View Full Product Details