Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exoc8
Entrez Gene ID
568029
Gene Name
exocyst complex component 8
Gene Aliases
zgc:162287
Chromosome Location
Chr.20: 4051752 - 4054046 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110475.1 NP_001103945.1 1 - 1 2045 61
GenBank mRNA BC134948.1 - 1 - 1 2045 61

View Full Product Details