Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wnt2
Entrez Gene ID
30127
Gene Name
wingless-type MMTV integration site family member 2
Gene Aliases
ZfWnt2, wnt-2
Chromosome Location
Chr.18: 211458 - 234686 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130950.1 NP_571025.1 4 - 5 1013 80
GenBank mRNA BC056277.1 - 4 - 5 1028 80
U51266.1 - 4 - 5 1013 80

View Full Product Details