Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dla
Entrez Gene ID
30131
Gene Name
deltaA
Gene Aliases
cb335, fa04c10, wu:fa04c10
Chromosome Location
Chr.1: 54013455 - 54026180 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130954.2 NP_571029.2 5 - 6 1152 111
GenBank mRNA AF030031.1 - 5 - 6 1143 111
BC075742.1 - 5 - 6 1152 111
CU458989.1 - 5 - 6 1143 111
DQ993185.1 - 5 - 6 1143 111

View Full Product Details