Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dla
Entrez Gene ID
30131
Gene Name
deltaA
Gene Aliases
cb335, fa04c10, wu:fa04c10
Chromosome Location
Chr.1: 54013455 - 54026180 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130954.2 NP_571029.2 9 - 10 2490 96
GenBank mRNA AF030031.1 - 9 - 10 2481 96
BC075742.1 - 9 - 10 2490 96
CU458989.1 - 9 - 10 2481 96
DQ993185.1 - 9 - 10 2481 96

View Full Product Details