Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ddx19
Entrez Gene ID
192339
Gene Name
DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (DBP5 homolog, yeast)
Gene Aliases
chunp6915, fi21a04, wu:fb24g07, wu:fc59e08, wu:fd14f03, wu:fi21a04
Chromosome Location
Chr.11: 3524737 - 3535500 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173258.1 NP_775365.1 7 - 8 764 80
GenBank mRNA AY099536.1 - 7 - 8 764 80
BC044541.1 - 7 - 8 80

View Full Product Details